Tisk…

Formaldehyde ≥34,5% in aqueous solution stabilised USP, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: 9.0-15.0 % Stabilised with methanol
M-Clarity™ quality level = MQ200

15512-1L-R 15512-2.5L-R 15512-25KG-R 15512-5L-R
SIAL15512-1L-REA 1070 CZK
SIAL15512-1L-R SIAL15512-2.5L-R SIAL15512-25KG-R SIAL15512-5L-R
Formaldehyde ≥34,5% in aqueous solution stabilised USP, Sigma-Aldrich®
Formaldehyde
Vzorec: CH₂O
MW: 30.03
Bod varu: -19.3
Bod tání: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS číslo: 50-00-0
UN: 2209
ADR: 8 III
Merck Index: 13,04259

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR