Tisk…

N,O-Bis(trimethylsilyl)trifluoroacetamide, Supelco®

Dodavatel: Merck
Nebezpečí

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

M-Clarity™ quality level = MQ100

15222-1ML-F 15222-25ML-F 15222-5ML-F 15222-10X1ML-F
15222-1ML-FEA 926 CZK
15222-1ML-F 15222-25ML-F 15222-5ML-F 15222-10X1ML-F
N,O-Bis(trimethylsilyl)trifluoroacetamide, Supelco®
N,O-Bis(trimethylsilyl)trifluoroacetamide
Vzorec: C₈H₁₈F₃NOSi₂
Bod varu: 145 °C (1013 hPa)
Bod tání: –41 °C
Hustota: 0,96 g/cm³ (20 °C)
Bod vzplanutí: 34 °C
MDL Number: MFCD00008269
CAS číslo: 25561-30-2
EINECS: 247-103-9
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08d8a2bd736dc46554a781803537a4d7f74cf5e8&vid=f2dc5486a30b1553dbc910bb90b948e1c110a4e0&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR