Tisk…

α-Bromo-p-toluonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(Bromomethyl)benzonitrile, 4-Cyanobenzyl bromide, p-(Bromomethyl)benzonitrile

M-Clarity™ quality level = MQ200

144061-10G 144061-1G 144061-50G
SIAL144061-10GEA 3540 CZK
SIAL144061-10G SIAL144061-1G SIAL144061-50G
α-Bromo-p-toluonitrile, Sigma-Aldrich®
α-Bromo-p-toluonitrile
Vzorec: C₈H₆BrN
MW: 196,05 g/mol
Bod varu: 135…137 °C (0,6 mmHg)
Bod tání: 115 °C
MDL Number: MFCD00001829
CAS číslo: 17201-43-3
EINECS: 241-246-0
UN: 2923
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR