Tisk…

N,N,N-Trimethylanilinium tribromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

M-Clarity™ quality level = MQ100

139718-100G 139718-25G
SIAL139718-100GEA 4740 CZK
SIAL139718-100G SIAL139718-25G
N,N,N-Trimethylanilinium tribromide, Sigma-Aldrich®
N,N,N-Trimethylanilinium tribromide
Vzorec: C₉H₁₄Br₃N
MW: 375,93 g/mol
Bod tání: 115…116 °C
MDL Number: MFCD00011789
CAS číslo: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR