Tisk…

Methyl crotonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl trans-2-butenoate, 2-Butenoic acid methyl ester, trans-Methyl crotonate, trans-Crotonic acid methyl ester, Crotonic acid methyl ester, trans-2-Butenoic acid methyl ester

M-Clarity™ quality level = MQ100

139459-100ML 139459-500ML 139459-5ML
SIAL139459-100MLEA 1270 CZK
SIAL139459-100ML SIAL139459-500ML SIAL139459-5ML
Methyl crotonate, Sigma-Aldrich®
Methyl crotonate
Vzorec: C₅H₈O₂
MW: 100,12 g/mol
Bod varu: 118 °C (1013 hPa)
Hustota: 0,95 g/cm³ (20 °C)
MDL Number: MFCD00009287
CAS číslo: 623-43-8
EINECS: 210-793-7
UN: 3272
ADR: 3,II
Merck Index: 13,02625

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR