Tisk…

2-Naphthalenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Naphthalene-2-sulphonyl chloride, β-Naphthalenesulphonyl chloride

M-Clarity™ quality level = MQ200

133361-25G 133361-5G
SIAL133361-25GEA 1840 CZK
SIAL133361-25G SIAL133361-5G
2-Naphthalenesulphonyl chloride, Sigma-Aldrich®
2-Naphthalenesulphonyl chloride
Vzorec: C₁₀H₇ClO₂S
Bod varu: 371 °C (1013 hPa)
Bod tání: 71…72 °C
MDL Number: MFCD00004087
CAS číslo: 93-11-8
EINECS: 202-219-9
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR