Indole-3-carbaldehyd, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde
M-Clarity™ quality level = MQ200
129445-25G
129445-5G
SIAL129445-25GEA
4310
CZK
SIAL129445-25G
SIAL129445-5G
Indole-3-carbaldehyd, Sigma-Aldrich®
Indole-3-carbaldehyd
Vzorec:
C₉H₇NO Bod tání: 192…195 °C Hustota: 1,34 g/cm³ (20 °C) |
MDL Number:
MFCD00005622 CAS číslo: 487-89-8 EINECS: 207-665-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...