1-Naphthylmethylamine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene
M-Clarity™ quality level = MQ100
127035-25G
127035-5G
SIAL127035-25GEA
8760
CZK
SIAL127035-25G
SIAL127035-5G
1-Naphthylmethylamine, Sigma-Aldrich®
1-Naphthylmethylamine
Vzorec:
C₁₁H₁₁N MW: 157,22 g/mol Bod varu: 92 °C (0,1 mmHg) Bod tání: 267…269 °C Hustota: 1,0958 g/cm³ (20 °C) |
MDL Number:
MFCD00004048 CAS číslo: 118-31-0 EINECS: 204-244-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...