Tisk…

5-Chlorovaleryl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Chlorovaleroyl chloride, 5-Chloropentanoyl chloride

M-Clarity™ quality level = MQ200

125245-10G 125245-50G
SIAL125245-10GEA 4310 CZK
SIAL125245-10G SIAL125245-50G
5-Chlorovaleryl chloride, Sigma-Aldrich®
5-Chlorovaleryl chloride
Vzorec: C₅H₈Cl₂O
MW: 155.02
Bod varu: 97…99 °C (1 mmHg)
Hustota: 1.21
MDL Number: MFCD00000758
CAS číslo: 1575-61-7
EINECS: 216-403-1
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR