tert-Butyl bromoacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Bromoacetic tert-butyl ester
M-Clarity™ quality level = MQ200
124230-10G
124230-250G
124230-50G
SIAL124230-10GEA
1080
CZK
SIAL124230-10G
SIAL124230-250G
SIAL124230-50G
tert-Butyl bromoacetate, Sigma-Aldrich®
tert-Butyl bromoacetate
Vzorec:
BrCH₂COOC(CH₃)₂ Bod varu: 50 °C (1013 hPa) Hustota: 1,338 g/cm³ (20 °C) |
MDL Number:
MFCD00000188 CAS číslo: 5292-43-3 EINECS: 226-133-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...