Tisk…

Ethyl levulinate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 4-oxopentanoate, 4-Oxovaleric acid ethyl ester, Ethyl 4-oxovalerate, 4-Ketovaleric acid ethyl ester, 4-Oxopentanoic acid ethyl ester, Levulinic acid ethyl ester

M-Clarity™ quality level = MQ200

122629-100G 122629-500G
SIAL122629-100GEA 1460 CZK
SIAL122629-100G SIAL122629-500G
Ethyl levulinate, Sigma-Aldrich®
Ethyl levulinate
Vzorec: C₇H₁₂O₃
MW: 144,17 g/mol
Bod varu: 93 °C (1013 hPa)
Hustota: 1,01 g/cm³ (20 °C)
MDL Number: MFCD00009209
CAS číslo: 539-88-8
EINECS: 208-728-2
Merck Index: 13,03852

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR