2-Formylbenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Carboxybenzaldehyde, Phthalaldehydic acid
M-Clarity™ quality level = MQ100
116017-100G
116017-25G
SIAL116017-100GEA
2180
CZK
SIAL116017-100G
SIAL116017-25G
2-Formylbenzoic acid, Sigma-Aldrich®
2-Formylbenzoic acid
Vzorec:
C₈H₆O₃ MW: 150,13 g/mol Bod varu: 101 °C (0,4 mmHg) Bod tání: 95…98 °C Hustota: 1,404 g/cm³ (20 °C) |
MDL Number:
MFCD00003336 CAS číslo: 119-67-5 EINECS: 204-342-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...