Tisk…

Isophthalaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: m-Phthalaldehyde, Isophthaldicarboxaldehyde, Isophthaldialdehyde

M-Clarity™ quality level = MQ200

115282-25G 115282-5G
SIAL115282-25GEA 7570 CZK
SIAL115282-25G SIAL115282-5G
Isophthalaldehyde, Sigma-Aldrich®
Isophthalaldehyde
Vzorec: C₈H₆O₂
MW: 134,13 g/mol
Bod varu: 255 °C (1013 hPa)
Bod tání: 89…91 °C
MDL Number: MFCD00003372
CAS číslo: 626-19-7
EINECS: 210-935-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR