Propyl propionate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
112267-100ML
112267-1L
SIAL112267-100MLEA
1680
CZK
SIAL112267-100ML
SIAL112267-1L
Propyl propionate, Sigma-Aldrich®
Propyl propionate
Vzorec:
C₆H₁₂O₂ Hustota: 0,88 g/cm³ (20 °C) |
MDL Number:
MFCD00009373 CAS číslo: 106-36-5 EINECS: 203-389-7 UN: 3272 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...