Tisk…

1,6-Heptadien-4-ol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

111589-25G 111589-5G
SIAL111589-25GEA 17160 CZK
SIAL111589-25G SIAL111589-5G
1,6-Heptadien-4-ol, Sigma-Aldrich®
1,6-Heptadien-4-ol
Vzorec: C₇H₁₂O
MW: 112.17
Hustota: 0.918
MDL Number: MFCD00008663
CAS číslo: 2883-45-6
EINECS: 220-742-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR