Tisk…

Butyl vinyl ether, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: n-Butyl vinyl ether

M-Clarity™ quality level = MQ200

110299-100ML 110299-1L 110299-2L 110299-500ML
SIAL110299-100MLEA 1920 CZK
SIAL110299-100ML SIAL110299-1L SIAL110299-2L SIAL110299-500ML
Butyl vinyl ether, Sigma-Aldrich®
Butyl vinyl ether
Vzorec: C₆H₁₂O
MW: 100,16 g/mol
Bod varu: 94 °C (1013 hPa)
Bod tání: –91,9 °C
Hustota: 0,7804 g/cm³ (20 °C)
MDL Number: MFCD00009454
CAS číslo: 111-34-2
EINECS: 203-860-7
UN: 3271
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR