Diphenyl phthalate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Phthalic acid diphenyl ester, Phenyl phthalate
M-Clarity™ quality level = MQ100
105880-100G
105880-25G
SIAL105880-100GEA
4150
CZK
SIAL105880-100G
SIAL105880-25G
Diphenyl phthalate, Sigma-Aldrich®
Diphenyl phthalate
Vzorec:
C₂₀H₁₄O₄ Bod varu: 405 °C (1013 hPa) Bod tání: 74…76 °C Hustota: 1,313 g/cm³ (20 °C) |
MDL Number:
MFCD00003038 CAS číslo: 84-62-8 EINECS: 201-546-4 Merck Index: 12,07460 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...