1,6-Bis(dimethylamino)hexane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N,N,N',N'-Tetramethylhexamethylenediamine, N,N,N',N'-Tetramethyl-1,6-diaminohexane, N1,N1,N6,N6-Tetramethylhexane-1,6-diamine, N,N,N',N'-Tetramethyl-1,6-hexanediamine
M-Clarity™ quality level = MQ200
105139-100ML
105139-1L
SIAL105139-100MLEA
4790
CZK
SIAL105139-100ML
SIAL105139-1L
1,6-Bis(dimethylamino)hexane, Sigma-Aldrich®
1,6-Bis(dimethylamino)hexane
Vzorec:
C₁₀H₂₄N₂ MW: 172,31 g/mol Bod varu: 209 °C (1013 hPa) Bod tání: –46 °C Hustota: 0,8064 g/cm³ (15 °C) |
MDL Number:
MFCD00008339 CAS číslo: 111-18-2 EINECS: 203-842-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...