Benzothiazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Benzo[d]thiazole
M-Clarity™ quality level = MQ100
101338-100G
101338-500G
101338-5G
SIAL101338-100GEA
1230
CZK
SIAL101338-100G
SIAL101338-500G
SIAL101338-5G
Benzothiazole, Sigma-Aldrich®
Benzothiazole
Vzorec:
C₇H₅NS MW: 135,19 g/mol Bod varu: 231 °C (1013 hPa) Hustota: 1,246 g/cm³ (20 °C) |
MDL Number:
MFCD00005775 CAS číslo: 95-16-9 EINECS: 202-396-2 UN: 2810 ADR: 6.1,III Merck Index: 13,01108 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...