Tisk…

3-Iodobenzylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (3-Iodophenyl)methanamine

100064-1G 100064-5G
SIAL100064-1GEA 5220 CZK
SIAL100064-1G SIAL100064-5G
3-Iodobenzylamine, Sigma-Aldrich®
3-Iodobenzylamine
Vzorec: C₇H₈IN
MW: 233.05
Bod varu: 132 °C (8 mmHg)
Hustota: 1.748
MDL Number: MFCD00192227
CAS číslo: 696-40-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR