Tisk…

EDTA tripotassium salt dihydrate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Ethylenedinitrilotetra-acetic acid tripotassium salt dihydrate, Tripotassium hydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid tripotassium salt dihydrate, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Edetic acid tripotassium salt dihydrate

MQ100

03664-100G 03664-500G
SIAL03664-100GEA 6660 CZK
SIAL03664-100G SIAL03664-500G
EDTA tripotassium salt dihydrate, Sigma-Aldrich®
EDTA tripotassium salt dihydrate
Vzorec: C₁₀H₁₃K₃N₂O₈·2H₂O
MW: 442.55
Storage Temperature: Ambient
MDL Number: MFCD00149672
CAS číslo: 65501-24-8
EINECS: 241-543-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR