Acetonitril
Dodavatel: SCHARLAB
|
Nebezpečí
|
AC03711000
AC03712500
SCARAC03711000EA
0
CZK
SCARAC03711000
SCARAC03712500
Acetonitril
Acetonitril
Vzorec:
H₃CCN Bod varu: 81,6 °C (1013 hPa) Hustota: 0,782 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS číslo: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...