2-Bromo-4-fluoro-1-iodobenzene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Bromo-1-fluoro-4-iodobenzene, 2-Bromo-4-fluoroiodobenzene, 1-Bromo-5-fluoro-2-iodobenzene
L20144.06
L20144.03
L20144.06EA
1880
CZK
L20144.06
L20144.03
2-Bromo-4-fluoro-1-iodobenzene ≥98%
2-Bromo-4-fluoro-1-iodobenzene
Purity: 98+%
CAS No.: 202865-73-4
Molecular Formula: C6H3BrFI
Formula Weight: 300.90
Flash Point:
Vzorec:
C₆H₃BrFI MW: 300.9 Bod varu: 224…225 °C Hustota: 1.33 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142868 CAS číslo: 202865-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...