2-(2-Fluorophenyl)ethanol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Fluorophenethyl alcohol
L19998.06
L19998.14
L19998.06EA
1540
CZK
L19998.06
L19998.14
2-(2-Fluorophenyl)ethanol ≥99%
2-(2-Fluorophenyl)ethanol
Purity: 99%
CAS No.: 50919-06-7
Molecular Formula: C8H9FO
Formula Weight: 140.16
Flash Point: 98°(208°F)
Refractive Index: 1.5100
Boiling Point: 72°/1mm
MDL No.: MFCD00002887
Vzorec:
C₈H₉FO MW: 140,16 g/mol Bod varu: 72 °C (1 mmHg) Hustota: 1,045 Bod vzplanutí: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002887 CAS číslo: 50919-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...