1-Ethyl-3-methylimidazoliumhexafluorophosphate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Ethyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)
L19762.04
L19762.09
L19762.18
L19762.04EA
1740
CZK
L19762.04
L19762.09
L19762.18
1-Ethyl-3-methylimidazoliumhexafluorophosphate ≥98%
1-Ethyl-3-methylimidazoliumhexafluorophosphate
Purity: 98+%
CAS No.: 155371-19-0
Molecular Formula: C6H11F6N2P
Formula Weight: 256.13
Melting Point: 58-62°
MDL No.: MFCD00216703
Vzorec:
C₆H₁₁F₆N₂P MW: 256,13 g/mol Bod tání: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216703 CAS číslo: 155371-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...