Tisk…

5-Methoxyindol-3-ylacetic acid ≥98%

Dodavatel: Thermo Fisher Scientific

L19073.03 L19073.MD
L19073.03EA 4020 CZK
L19073.03 L19073.MD
5-Methoxyindol-3-ylacetic acid ≥98%
5-Methoxyindol-3-ylacetic acid

Purity: 98+% CAS No.: 3471-31-6 Molecular Formula: C11H11NO3 Formula Weight: 205.22 Melting Point: 145-148° dec. Beilstein Reference: 187161

Vzorec: C₁₁H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00005638
CAS číslo: 3471-31-6
EINECS: 222-438-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR