Allyl isocyanate 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Isocyanatoprop-1-ene
L18710.03
L18710.06
L18710.03EA
1400
CZK
L18710.03
L18710.06
Allyl isocyanate 96%
Allyl isocyanate
Purity: 98%
CAS No.: 1476-23-9
Molecular Formula: CH2=CHCH2NCO
Formula Weight: 83.09
Storage Temperature: 2°C to 8°C
Flash Point: 6°(43°F)
Refractive Index: 1.4185
Odour: Pungent
Boiling Point: 87-89°
Beilstein Reference: 506106
UN No.: 2478
MDL No.: MFCD00002044
Vzorec:
C₄H₅NO MW: 83,09 g/mol Bod varu: 87…89 °C Hustota: 0,940 Bod vzplanutí: 6 °C (43 °F) |
MDL Number:
MFCD00002044 CAS číslo: 1476-23-9 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...