3-(Dimethylamino)benzoyl chloride hydrochloride tech. 90%, Technical Grade
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L18433.06
L18433.03
L18433.06EA
8930
CZK
L18433.06
L18433.03
3-(Dimethylamino)benzoyl chloride hydrochloride tech. 90%, Technical Grade
3-(Dimethylamino)benzoyl chloride hydrochloride
Purity: 90%
CAS No.: 117500-61-5
Molecular Formula: C9H10ClNO.HCl
Formula Weight: 220.10
Melting Point: 131-133°
Beilstein Reference: 2803337
UN No.: 3261
MDL No.: MFCD02094023
Vzorec:
C₉H₁₁Cl₂NO Bod tání: 131…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD19442144 CAS číslo: 117500-61-5 UN: 3261 ADR: 8,II |