Dimethyl azelate ≥97%
Dodavatel: Thermo Scientific
Synonyms:
1,9-Nonanedioic acid dimethyl ester, Dimethyl nonanedioate, Azelaic acid dimethyl ester
L17458.06
L17458.14
L17458.22
L17458.06EA
1400
CZK
L17458.06
L17458.14
L17458.22
Dimethyl azelate ≥97%
Dimethyl azelate
Purity: 97%
CAS No.: 1732-10-1
Molecular Formula: C11H20O4
Formula Weight: 216.28
Flash Point:
Vzorec:
C₁₁H₂₀O₄ MW: 216,28 g/mol Bod varu: 156 °C (20 mmHg) Bod tání: 18 °C Hustota: 1,007 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025898 CAS číslo: 1732-10-1 EINECS: 217-060-0 Merck Index: 14,905 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...