6-(Trifluoromethyl)indole ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
L17009.03
L17009.MD
L17009.03EA
3180
CZK
L17009.03
L17009.MD
6-(Trifluoromethyl)indole ≥97%
6-(Trifluoromethyl)indole
Purity: 97%
CAS No.: 13544-43-9
Molecular Formula: C9H6F3N
Formula Weight: 185.15
Melting Point: 101-105°
Beilstein Reference: 1242785
Vzorec:
C₉H₆F₃N MW: 185,15 g/mol Bod tání: 101…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272316 CAS číslo: 13544-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...