Vinyltrimethylsilane ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98+%
CAS No.: 754-05-2
Molecular Formula: C5H12Si
Formula Weight: 100.24
Flash Point: -24°(-11°F)
Refractive Index: 1.3910
Odour: Mild
Melting Point: -132°
Boiling Point: 54-56°
Beilstein Reference: 956572
UN No.: 1993
MDL No.: MFCD00008606
Vzorec:
C₅H₁₂Si Bod varu: 54…56 °C Bod tání: –132 °C Hustota: 0,690 Bod vzplanutí: –24 °C (–11 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008606 CAS číslo: 754-05-2 EINECS: 212-042-9 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...