2,3-Dimethylfuran ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L15705.03
L15705.06
L15705.03EA
1340
CZK
L15705.03
L15705.06
2,3-Dimethylfuran ≥99%
2,3-Dimethylfuran
Purity: 99%
CAS No.: 14920-89-9
Molecular Formula: C6H8O
Formula Weight: 96.13
Flash Point: -1°(30°F)
Refractive Index: 1.4430
Boiling Point: 42°/15mm
Beilstein Reference: 104572
UN No.: 1992
MDL No.: MFCD00153893
Vzorec:
C₆H₈O MW: 96,13 g/mol Bod varu: 42 °C (115 mmHg) Hustota: 0,910 Bod vzplanutí: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00153893 CAS číslo: 14920-89-9 UN: 1992 ADR: 3,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...