Tisk…

3-(2-Chlorophenyl)propionic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-(2-Chlorophenyl)propanoic acid

L15306.03 L15306.06
L15306.03EA 1890 CZK
L15306.03 L15306.06
3-(2-Chlorophenyl)propionic acid ≥98%
3-(2-Chlorophenyl)propionic acid

Purity: 98+% CAS No.: 1643-28-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 94-97° Boiling Point: 171°/10mm MDL No.: MFCD00016547

Vzorec: C₉H₉ClO₂
MW: 184.62
Bod varu: 171 °C (10 mmHg)
Bod tání: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00016547
CAS číslo: 1643-28-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR