Isobutyronitrile ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 99%
CAS No.: 78-82-0
Molecular Formula: C4H7N
Formula Weight: 69.11
Flash Point: 8°(46°F)
Refractive Index: 1.3740
Melting Point: -72°
Boiling Point: 103-104°
Beilstein Reference: 1340512
Merck Reference: 14,5156
UN No.: 2284
MDL No.: MFCD00001873
Vzorec:
C₄H₇N Bod varu: 103…104 °C Bod tání: –72 °C Hustota: 0,770 Bod vzplanutí: 8 °C (46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001873 CAS číslo: 78-82-0 EINECS: 201-147-5 UN: 2284 ADR: 3,II Merck Index: 14,5156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...