Tisk…

1-Chloroethyl chloroformate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-Chloroethyl carbonochloridate

L14197.06 L14197.14
L14197.06EA 951 CZK
L14197.06 L14197.14
1-Chloroethyl chloroformate ≥98%
1-Chloroethyl chloroformate

Purity: 98% CAS No.: 50893-53-3 Molecular Formula: C3H4Cl2O2 Formula Weight: 142.97 Storage Temperature: 2°C to 8°C Flash Point: 45°(113°F) Refractive Index: 1.4230 Melting Point: -65° Boiling Point: 119-121° Beilstein Reference: 1747601 UN No.: 2742 MDL No.: MFCD00000647

Vzorec: C₃H₄Cl₂O₂
MW: 142.97
Bod varu: 119…121 °C
Bod tání: –65 °C
Hustota: 1.312
Bod vzplanutí: 45 °C (113 °F)
MDL Number: MFCD00000647
CAS číslo: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR