N-Methylcyclohexylamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
N-Cyclohexylmethylamine, Cyclohexyl(methyl)amine
L13872.AC
L13872.AE
L13872.ACEA
646
CZK
L13872.AC
L13872.AE
N-Methylcyclohexylamine 98%
N-Methylcyclohexylamine
Purity: 98%
CAS No.: 100-60-7
Molecular Formula: C7H15N
Formula Weight: 113.20
Flash Point: 35°(95°F)
Refractive Index: 1.4560
Melting Point: -8°
Boiling Point: 149°
Beilstein Reference: 1523664
UN No.: 2734
MDL No.: MFCD00003832
Vzorec:
C₇H₁₅N MW: 113,2 g/mol Bod varu: 149 °C Bod tání: –8 °C Hustota: 0,857 Bod vzplanutí: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003832 CAS číslo: 100-60-7 EINECS: 202-869-3 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...