3,4-Dibromobenzaldehyde ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
L13762.03
L13762.06
L13762.03EA
2940
CZK
L13762.03
L13762.06
3,4-Dibromobenzaldehyde ≥99%
3,4-Dibromobenzaldehyde
Purity: 99%
CAS No.: 74003-55-7
Molecular Formula: C7H4Br2O
Formula Weight: 263.93
Melting Point: 69-72°
Beilstein Reference: 2573433
Vzorec:
C₇H₄Br₂O MW: 263,92 g/mol Bod tání: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236033 CAS číslo: 74003-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...