Tisk…

3,4,5-Trimethoxybenzylamine 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L13461.06 L13461.14
L13461.06EA 2160 CZK
L13461.06 L13461.14
3,4,5-Trimethoxybenzylamine 96%
3,4,5-Trimethoxybenzylamine

Purity: 96% CAS No.: 18638-99-8 Molecular Formula: C10H15NO3 Formula Weight: 197.24 Flash Point: 110°(230°F) Refractive Index: 1.5480 Boiling Point: 120-122°/0.05m Beilstein Reference: 1107570 UN No.: 2735 MDL No.: MFCD00017150

Vzorec: C₁₀H₁₅NO₃
Bod varu: 120…122 °C (0,05 m)
Hustota: 1,155
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017150
CAS číslo: 18638-99-8
EINECS: 242-468-0
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR