Tisk…

Isobutyric anhydride ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Methylpropanoic anhydride, 2-Methylpropionic anhydride

L13240.AE L13240.AP L13240.0F
L13240.AEEA 848 CZK
L13240.AE L13240.AP L13240.0F
Isobutyric anhydride ≥97%
Isobutyric anhydride

Purity: 97% CAS No.: 97-72-3 Molecular Formula: [(CH3)2CHCO]2O Formula Weight: 158.20 Flash Point: 67°(152°F) Refractive Index: 1.4060 Odour: Pungent Melting Point: -56° Boiling Point: 182° Beilstein Reference: 386267 UN No.: 2922 MDL No.: MFCD00008913

Vzorec: C₈H₁₄O₃
Bod varu: 182 °C
Bod tání: –56 °C
Hustota: 0,954
Bod vzplanutí: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008913
CAS číslo: 97-72-3
EINECS: 202-603-6
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR