Tisk…

1-(4-Hydroxyphenyl)-2-thiourea ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: N-(4-Hydroxyphenyl)thiourea, 4-Hydroxyphenylthiourea

L12537.03 L12537.06
L12537.03EA 1190 CZK
L12537.03 L12537.06
1-(4-Hydroxyphenyl)-2-thiourea ≥98%
1-(4-Hydroxyphenyl)-2-thiourea

Purity: 98+% CAS No.: 1520-27-0 Molecular Formula: C7H8N2OS Formula Weight: 168.22 Odour: Odourless Melting Point: ca 210° dec. Beilstein Reference: 1102429 MDL No.: MFCD00041186

Vzorec: (C₆H₄(OH))NC(S)NH₂
MW: 168,22 g/mol
Bod tání: ∼ 210 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00041186
CAS číslo: 1520-27-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR