Tisk…

EDTA tripotassium salt dihydrate 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ethylenedinitrilotetra-acetic acid tripotassium salt dihydrate, Tripotassium hydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid tripotassium salt dihydrate, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Edetic acid tripotassium salt dihydrate

L11991.18 L11991.30
L11991.18EA 1030 CZK
L11991.18 L11991.30
EDTA tripotassium salt dihydrate 99%
EDTA tripotassium salt dihydrate

Purity: 99% CAS No.: 65501-24-8 Molecular Formula: C10H13K3N2O8.2H2O Formula Weight: 442.55 (406.46anhy) Melting Point: ca 182° dec. Beilstein Reference: 6762497

Vzorec: C₁₀H₁₃K₃N₂O₈·2H₂O
MW: 442.55
Bod tání: ∼ 182 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00149672
CAS číslo: 65501-24-8
EINECS: 241-543-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR