2,2'-butyliminodiethanol 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
N-Butyldiethanolamine, N,N-Diethanolbutylamine, N-N-Butyldiethanolamine, N-Butyl-2,2'-iminodiethanol
L09953.22
L09953.36
L09953.22EA
628
CZK
L09953.22
L09953.36
2,2'-butyliminodiethanol 98%
2,2'-butyliminodiethanol
Purity: 98%
CAS No.: 102-79-4
Molecular Formula: CH3(CH2)3N(CH2CH2OH)2
Formula Weight: 161.25
Flash Point: 143°(289°F)
Refractive Index: 1.4640
Melting Point: -70°
Boiling Point: 268-272°
Beilstein Reference: 1739642
UN No.: 2735
MDL No.: MFCD00002856
Vzorec:
H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂ MW: 161,24 g/mol Bod varu: 268…272 °C Bod tání: –70 °C Hustota: 0,968 Bod vzplanutí: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002856 CAS číslo: 102-79-4 EINECS: 203-055-0 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...