Tisk…

4-Propoxybenzaldehyde ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L08822.14 L08822.06
L08822.14EA 3400 CZK
L08822.14 L08822.06
4-Propoxybenzaldehyde ≥97%
4-Propoxybenzaldehyde

Purity: 97% CAS No.: 5736-85-6 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5480 Boiling Point: 129-130°/9mm Beilstein Reference: 743408 MDL No.: MFCD00014134

Vzorec: C₁₀H₁₂O₂
MW: 164.2
Bod varu: 129…130 °C (9 mmHg)
Hustota: 1.039
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014134
CAS číslo: 5736-85-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR