Tisk…

7-Azaindole ≥98%

Dodavatel: Thermo Fisher Scientific

L07983.03 L07983.06 L07983.14
L07983.03EA 1270 CZK
L07983.03 L07983.06 L07983.14
7-Azaindole ≥98%
7-Azaindole

Purity: 98% CAS No.: 271-63-6 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 103-107° Boiling Point: 270° MDL No.: MFCD00005606

Vzorec: C₇H₆N₂
Bod varu: 270 °C
Bod tání: 103…107 °C
Hustota: 1,301 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005606
CAS číslo: 271-63-6
EINECS: 205-981-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR