2-Chlorobenzoxazole 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Chloro-1,3-benzoxazole
L07981.06
L07981.14
L07981.06EA
1570
CZK
L07981.06
L07981.14
2-Chlorobenzoxazole 98%
2-Chlorobenzoxazole
Purity: 99%
CAS No.: 615-18-9
Molecular Formula: C7H4ClNO
Formula Weight: 153.57
Storage Temperature: 2°C to 8°C
Flash Point: 85°(185°F)
Refractive Index: 1.5660
Boiling Point: 201-202°
Beilstein Reference: 115982
MDL No.: MFCD00005766
Vzorec:
C₇H₄ClNO MW: 153.57 Bod varu: 201…202 °C Bod tání: 45 Hustota: 1.3453 Bod vzplanutí: 85 °C (185 °F) |
MDL Number:
MFCD00005766 CAS číslo: 615-18-9 EINECS: 210-414-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...