3,4-Dimethyl-o-phenylenediamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-3,4-dimethylphenylamine, 3,4-Dimethyl-1,2-phenylenediamine, 3,4-Dimethylbenzene-1,2-diamine, 1,2-Diamino-3,4-dimethylbenzene
Purity: 98%
CAS No.: 41927-01-9
Molecular Formula: C8H12N2
Formula Weight: 136.20
Melting Point: 86-90°
Beilstein Reference: 2716459
UN No.: 2811
MDL No.: MFCD00051709
Vzorec:
C₈H₁₂N₂ MW: 136,2 g/mol Bod varu: 287 °C (1013 hPa) Bod tání: 86…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051709 CAS číslo: 41927-01-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...