4-Bromopyridinium chloride ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Bromopyridine HCl, 4-Bromopyridine hydrochloride
L07052.06
L07052.14
L07052.06EA
2280
CZK
L07052.06
L07052.14
4-Bromopyridinium chloride ≥99%
4-Bromopyridinium chloride
Purity: 99%
CAS No.: 19524-06-2
Molecular Formula: C5H4BrN.HCl
Formula Weight: 194.46
Melting Point: ca 270° dec.
Beilstein Reference: 3621847
Vzorec:
C₅H₅BrClN MW: 194,46 g/mol Bod tání: ∼ 270 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012828 CAS číslo: 19524-06-2 EINECS: 243-128-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...