5-Amino-1H-indazole 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine
L06705.06
L06705.14
L06705.06EA
1620
CZK
L06705.06
L06705.14
5-Amino-1H-indazole 98%
5-Amino-1H-indazole
Purity: 98%
CAS No.: 19335-11-6
Molecular Formula: C7H7N3
Formula Weight: 133.15
Odour: Odourless
Melting Point: 172-178°
Beilstein Reference: 3259
UN No.: 2811
MDL No.: MFCD00037975
Vzorec:
C₇H₇N₃ MW: 133,15 g/mol Bod varu: 345 °C (1013 hPa) Bod tání: 172…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037975 CAS číslo: 19335-11-6 EINECS: 242-971-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...