2,2'-Dichlorobenzophenone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,2-DBP, Bis(2-chlorophenyl) ketone
L06144.03
L06144.06
L06144.03EA
2250
CZK
L06144.03
L06144.06
2,2'-Dichlorobenzophenone ≥98%
2,2'-Dichlorobenzophenone
Purity: 98+%
CAS No.: 5293-97-0
Molecular Formula: C13H8Cl2O
Formula Weight: 251.11
Melting Point: 50-53°
Beilstein Reference: 2050630
Vzorec:
C₁₃H₈Cl₂O MW: 251,11 g/mol Bod tání: 50…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039303 CAS číslo: 5293-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...