Decyltrichlorosilane ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Trichloro(decyl)silane
L05912.09
L05912.18
L05912.09EA
1410
CZK
L05912.09
L05912.18
Decyltrichlorosilane ≥97%
Decyltrichlorosilane
Purity: 97%
CAS No.: 13829-21-5
Molecular Formula: C10H21Cl3Si
Formula Weight: 275.72
Flash Point:
Vzorec:
C₁₀H₂₁Cl₃Si MW: 275,72 g/mol Bod varu: 133…137 °C (5 mmHg) Hustota: 1,049 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013600 CAS číslo: 13829-21-5 EINECS: 237-540-3 UN: 2987 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...